Learn More
2-Nitrobenzaldehyde, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 2-Nitrobenzaldehyde |
| CAS | 552-89-6 |
| Melting Point | 43.5°C |
| Boiling Point | 156°C (20 hPa) |
| Molecular Formula | 2-(NO2)C6H4CHO |
| Quantity | 1 g |
| Synonym | 2-Nitrobenzaldehyde, 2-Nitrobenzaldehyde |
| Molecular Weight (g/mol) | 151.12 |
| Grade | Not Available |