Få mere at vide
2-Nitroaniline-3,4,5,6-d4, CDN
Artikelnummer. 30323459
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323459 0.05 g -
30319031 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323459 Leverandør CDN Leverandørnr. CDND54370.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitroaniline-3,4,5,6-D4
CAS 204244-80-4
Molekylær formel C6 D4 H2 N2 O2
Mængde 0.05 g
Synonym Benzen-2,3,4,5-d4-amine, 6-nitro- (9 CI), 2-Nitrobenzenamine-3,4,5,6-d4, 1-Amino-2-nitrobenzene-3,4,5,6-d4, 1-Nitro-2-aminobenzene-3,4,5,6-d4, 2-Aminonitrobenzene-3,4,5,6-d4, 2-Nitroaniline-d4, 2-Nitroaniline-3,4,5,6-D4, o-Aminonitrobenzene-d4, o-Nitroaniline-d4
InChI formel InChI=1 S/C6H6N2O2/c7-5-3-1-2-4-6(5)8(9)10/h1-4 H,7H2/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(c(N)c1[2 H])[N+](=O)[O-]
IUPAC navn 2,3,4,5-tetradeuterio-6-nitroaniline
Molekylvægt (g/mol) 142.1487
Formel vægt 142.068 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity