missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-5-(1-piperidinyl)phenol, 97%, Thermo Scientific™

Catalog Number 11411347
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
157831-75-9
C11H13N2O3
221.24
MFCD00052663
DDKAQSBPHIORAR-UHFFFAOYSA-M
2-nitro-5-piperidinophenol, 2-nitro-5-piperidin-1-yl phenol, 2-nitro-5-1-piperidinyl phenol, phenol,2-nitro-5-1-piperidinyl, maybridge1_001384, acmc-20anp3, 2-nitro-5-piperidylphenol
2774482
2-nitro-5-piperidin-1-ylphenol
[O-]C1=CC(=CC=C1[N+]([O-])=O)N1CCCCC1
This item is not returnable. View return policy
157831-75-9
C11H13N2O3
221.24
MFCD00052663
DDKAQSBPHIORAR-UHFFFAOYSA-M
2-nitro-5-piperidinophenol, 2-nitro-5-piperidin-1-yl phenol, 2-nitro-5-1-piperidinyl phenol, phenol,2-nitro-5-1-piperidinyl, maybridge1_001384, acmc-20anp3, 2-nitro-5-piperidylphenol
2774482
2-nitro-5-piperidin-1-ylphenol
[O-]C1=CC(=CC=C1[N+]([O-])=O)N1CCCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 70°C
Quantity 1 g
Formula Weight 224.24
Percent Purity 97%
Chemical Name or Material 2-Nitro-5-(1-piperidinyl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'