Få mere at vide
2-Nitro-3-pentanol, TRC
Artikelnummer. 30482264
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482264 1 g
30479227 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482264 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4969101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitro-3-pentanol
CAS 20575-40-0
Molekylær formel C5 H11 N O3
Mængde 1 g
InChI formel InChI=1S/C5H11NO3/c1-3-5(7)4(2)6(8)9/h4-5,7H,3H2,1-2H3
SMIL CCC(O)C(C)[N+](=O)[O-]
IUPAC navn 2-nitropentan-3-ol
Molekylvægt (g/mol) 133.15
Formel vægt 133.0739