Få mere at vide
2-NBDG, TRC
Artikelnummer. 30480592
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30480592 2.5 mg 2.5mg
30479612 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480592 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3812982.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 186689-07-6
Molekylær formel C12 H14 N4 O8
Molekylvægt (g/mol) 342.26
Synonym 2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, NBD-Glucose, 2-NBDG
IUPAC navn (3R,4R,5S,6R)-6-(hydroxymethyl)-3-[(4-nitro-2,1,3-benzoxadiazol-7-yl)amino]tetrahydropyran-2,4,5-triol
SMIL OC[C@H]1OC(O)[C@H](Nc2ccc(c3nonc23)[N+](=O)[O-])[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale 2-NBD Glucose
Mængde 2.5 mg
InChI formel InChI=1S/C12H14N4O8/c17-3-6-10(18)11(19)9(12(20)23-6)13-4-1-2-5(16(21)22)8-7(4)14-24-15-8/h1-2,6,9-13,17-20H,3H2/t6-,9-,10-,11-,12?/m1/s1
Formel vægt 342.0812