Få mere at vide
2-Naphthylamine Hydrochloride, TRC
Artikelnummer. 30469820
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469820 10 mg
30481145 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469820 Leverandør Toronto Research Chemicals Leverandørnr. TRCN37802010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 612-52-2
Molekylær formel C10H9N . (HCl)
Molekylvægt (g/mol) 143.18
Synonym 2-Naphthalenamine Hydrochloride, 2-Aminonaphthalene Hydrochloride, β-Naphthylamine Hydrochloride
IUPAC navn naphthalen-2-amine;hydrochloride
SMIL Cl.Nc1ccc2ccccc2c1
Tekniske data
Kemisk navn eller materiale 2-Naphthylamine Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C10H9N/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,11H2
Formel vægt 179.0502