Få mere at vide
2-Naphthol-d8, TRC
Artikelnummer. 30482471
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30482471 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482471 Leverandør Toronto Research Chemicals Leverandørnr. TRCN36801650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Naphthol-d8
CAS 78832-61-8
Molekylær formel C10 D8 O
Mængde 50 mg
Synonym 2-Naphthalen-1,3,4,5,6,7,8-d7-ol-d (9CI), O,1,3,4,5,6,7,8-Octadeuterionaphthalen-2-ol, 2-Naphthol-D8, perdeutero-2-Naphthol, 2-Naphthol-O,1,3,4,5,6,7,8-d8, β-Naphthol-d8
InChI formel InChI=1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H/i1D,2D,3D,4D,5D,6D,7D/hD
SMIL [2H]Oc1c([2H])c([2H])c2c([2H])c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn 1,2,3,4,5,6,8-heptadeuterio-7-deuteriooxynaphthalene
Molekylvægt (g/mol) 152.2192
Formel vægt 152.1077