Få mere at vide
2-Naphthol-d7, TRC
Artikelnummer. 30469019
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30469019 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469019 Leverandør Toronto Research Chemicals Leverandørnr. TRCN36801825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Naphthol (ring-d7)
CAS 78832-54-9
Renhedsgrad noter HPLC
Molekylær formel C10 2H7 H O
Mængde 25 mg
Synonym 2-Naphthalen-1,3,4,5,6,7,8-d7-ol, 2-Naphthol-d7, 2-Naphthol (ring-D7)
InChI formel InChI=1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c2c([2H])c(O)c([2H])c([2H])c2c1[2H]
IUPAC navn 1,3,4,5,6,7,8-heptadeuterionaphthalen-2-ol
Molekylvægt (g/mol) 151.21
Formel vægt 151.1015
Procent renhed >95