missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
2-Naphthalenecarboxylic Acid-13C6, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 2-Naphthalenecarboxylic Acid-13C6 |
| CAS | 1346602-42-3 |
| Molekylær formel | C513C6H8O2 |
| Mængde | 2.5 mg |
| Synonym | 2-Naphthoic Acid-13C6, 2-Carboxynaphthalene-13C6, 2-Maythic Acid-13C6, 2-Naphthylcarboxylic Acid-13C6, Isonaphthoic Acid-13C6, beta-Naphthalenecarboxylic Acid-13C6, beta-Naphthoic Acid-13C6 |
| InChI formel | InChI=1S/C11H8O2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,(H,12,13)/i1+1,2+1,3+1,4+1,8+1,9+1 |
| SMIL | OC(=O)c1cc[13c]2[13cH][13cH][13cH][13cH][13c]2c1 |
| IUPAC navn | naphthalene-2-carboxylic acid |
| Molekylvægt (g/mol) | 178.14 |
| Formel vægt | 178.0726 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?