Få mere at vide
2-(Naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole Hydrochloride (beta-Naphazoline Hydrochloride), Mikromol™
Beskrivelse
2-(Naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole Hydrochloride (beta-Naphazoline Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | beta-Naphazoline Hydrochloride |
| CAS | 6028-15-5 |
| Analyt- eller komponentnavne | beta-Naphazoline Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H14 N2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Naphazoline Hydrochloride Imp. D (EP) as Hydrochloride, 2-(Naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole Hydrochloride, beta-Naphazoline Hydrochloride, 4,5-Dihydro-2-(2-naphthalenylmethyl)-1H-imidazole hydrochloride, 4,5-Dihydro-2-(2-naphthalenylmethyl)-1H-imidazole monohydrochloride, 2-(2-Naphthylmethyl)-2-imidazoline hydrochloride, Naphazoline Hydrochloride Impurity D as Hydrochloride, Naphazoline Nitrate Impurity D as Hydrochloride, Naphazoline Impurity D as Hydrochloride, Naphazoline Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C14H14N2.ClH/c1-2-4-13-9-11(5-6-12(13)3-1)10-14-15-7-8-16-14;/h1-6,9H,7-8,10H2,(H,15,16);1H |
| SMIL | Cl.C(C1=NCCN1)c2ccc3ccccc3c2 |
| IUPAC navn | 2-(naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride |
| Vis mere |