Learn More
Alfa Aesar™ 2-n-Propylthiazolidine-4-carboxylic acid, 97%
Brand:  Alfa Aesar™ H62991.03
Additional Details : CAS Number : 4165-34-8 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469418
  • 1912.64 DKK / 1g
Estimated Shipment date
from Supplier 17-10-2018
Add to basket
Specifications
Chemical Identifiers
4165-34-8
175.246
QIJPWSGHURHDHJ-UHFFFAOYSA-N
2-propyl-1,3-thiazolidine-4-carboxylic acid
C7H13NO2S
MFCD09930114
165189
CCCC1NC(CS1)C(=O)O
Specifications
2-n-Propylthiazolidine-4-carboxylic acid
4165-34-8
C7H13NO2S
QIJPWSGHURHDHJ-UHFFFAOYSA-N
2-propyl-1,3-thiazolidine-4-carboxylic acid
165189
Solid
1g
97%
MFCD09930114
CCCC1NC(CS1)C(=O)O
175.246
175.25