Få mere at vide
2-N-Propyl Pramipexole-d4, TRC
Artikelnummer. 30419247
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30419247 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419247 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83600210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-N-Propyl Pramipexole-d4
CAS 1346602-54-7
Molekylær formel C13H19D4N3S
Mængde 10 mg
Synonym (S)-4,5,6,7-Tetrahydro-N2-propyl-N6-propyl-2,6-benzothiazolediamine-d4
InChI formel InChI=1S/C13H23N3S/c1-3-7-14-10-5-6-11-12(9-10)17-13(16-11)15-8-4-2/h10,14H,3-9H2,1-2H3,(H,15,16)/t10-/m0/s1/i7D2,8D2
SMIL [2H]C([2H])(CC)N[C@H]1CCc2nc(NC([2H])([2H])CC)sc2C1
IUPAC navn (6S)-2-N,6-N-bis(1,1-dideuteriopropyl)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Molekylvægt (g/mol) 257.43
Formel vægt 257.1864