Få mere at vide
2-n-Propyl-4-methyl-6-(1-methylbenzimidazol-2-yl)-benzimidazole-d3, TRC
Artikelnummer. 30421980
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30421980 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421980 Leverandør Toronto Research Chemicals Leverandørnr. TRCP838282100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-n-Propyl-4-methyl-6-(1-methylbenzimidazol-2-yl)-benzimidazole-d3
CAS 1185154-60-2
Molekylær formel C19H17D3N4
Mængde 100 mg
Synonym 2-n-Propyl-4-methyl-6-(1-methyl-d3-benzimidazol-2-yl)-benzimidazole, 4-Methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazole-d3
InChI formel InChI=1S/C19H20N4/c1-4-7-17-20-15-11-13(10-12(2)18(15)22-17)19-21-14-8-5-6-9-16(14)23(19)3/h5-6,8-11H,4,7H2,1-3H3,(H,20,22)/i3D3
SMIL [2H]C([2H])([2H])n1c(nc2ccccc12)c3cc(C)c4nc(CCC)[nH]c4c3
IUPAC navn 4-methyl-2-propyl-6-[1-(trideuteriomethyl)benzimidazol-2-yl]-1H-benzimidazole
Molekylvægt (g/mol) 307.41
Formel vægt 307.1876