Få mere at vide
2-(N-Methyl-2-pyrrolidinyl)ethyl Chloride Hydrochloride, TRC
Artikelnummer. 30469988
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469988 10 g
30473861 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469988 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32073510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56824-22-7
Molekylær formel C7 H14 Cl N . HCl
Molekylvægt (g/mol) 184.1
Synonym 2-(2-Chloroethyl)-1-methyl-pyrrolidine Hydrochloride, 1-Methyl-2-(2-chloroethyl)pyrrolidine Hydrochloride, 2-(2-Chloroethyl)-1-methylpyrrolidine Hydrochloride, 2-(2-Chloroethyl)-N-methylpyrrolidine Hydrochloride, N-Methyl-2-(2-chloroethyl)pyrrolidine Hydrochloride
IUPAC navn 2-(2-chloroethyl)-1-methylpyrrolidine hydrochloride
SMIL ClCCC1N(CCC1)C.Cl
Tekniske data
Kemisk navn eller materiale 2-(N-Methyl-2-pyrrolidinyl)ethyl Chloride Hydrochloride
Mængde 10 g
InChI formel InChI=1S/C7H14ClN.ClH/c1-9-6-2-3-7(9)4-5-8;/h7H,2-6H2,1H3;1H
Formel vægt 183.06