missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Hexyloxy)ethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7026405
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
112-25-4
C8H18O2
146.23
MFCD00045997
UPGSWASWQBLSKZ-UHFFFAOYSA-N
2-hexyloxy ethanol, ethanol, 2-hexyloxy, 2-hexyloxyethanol, ethylene glycol monohexyl ether, hexyl cellosolve, n-hexyl cellosolve, glycol monohexyl ether, cellosolve, n-hexyl, ethylene glycol n-hexyl ether
8170
CCCCCCOCCO
This item is not returnable. View return policy
112-25-4
C8H18O2
146.23
MFCD00045997
UPGSWASWQBLSKZ-UHFFFAOYSA-N
2-hexyloxy ethanol, ethanol, 2-hexyloxy, 2-hexyloxyethanol, ethylene glycol monohexyl ether, hexyl cellosolve, n-hexyl cellosolve, glycol monohexyl ether, cellosolve, n-hexyl, ethylene glycol n-hexyl ether
8170
CCCCCCOCCO

2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

Solubility
Soluble in alcohol and ether, water (9.46 g/L ).

Notes
Store under inert gas. Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point -42°C
Density 0.888
Boiling Point 208°C
Flash Point 94°C (201°F)
Linear Formula CH3(CH2)5OCH2CH2OH
Refractive Index 1.433
Quantity 1 g
UN Number UN2922
Beilstein 1734691
Solubility Information Soluble in alcohol and ether,water (9.46g/L ).
IUPAC Name 2-hexoxyethanol
Formula Weight 146.23
Percent Purity 99%
Chemical Name or Material 2-(n-Hexyloxy)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'