missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(N-Ethylanilino)ethanol, TRC

Artikelnummer. 30376771
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376771 5 g
30384607 25 g
30379757 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30376771 Leverandør Toronto Research Chemicals Leverandørnr. TRCE6787655G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 92-50-2
Molekylær formel C10H15NO
Molekylvægt (g/mol) 165.23
Synonym 2-(Ethylphenylamino)ethanol, Ethylanilinoethanol, Ethylphenylethanolamine, Hydroxyethylethylaniline, N-(2-Hydroxyethyl)-N-ethylaniline, Phenylethylethanolamine
IUPAC navn 2-(N-ethylanilino)ethanol
SMIL CCN(CCO)c1ccccc1

Tekniske data

Kemisk navn eller materiale 2-(N-Ethylanilino)ethanol
Mængde 5 g
InChI formel InChI=1S/C10H15NO/c1-2-11(8-9-12)10-6-4-3-5-7-10/h3-7,12H,2,8-9H2,1H3
Formel vægt 165.1154
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.