missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butylthiophene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020695
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1455-20-5
C8H12S
140.244
MFCD00022534
MNDZHERKKXUTOE-UHFFFAOYSA-N
2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak
73818
2-butylthiophene
CCCCC1=CC=CS1
This item is not returnable. View return policy
1455-20-5
C8H12S
140.244
MFCD00022534
MNDZHERKKXUTOE-UHFFFAOYSA-N
2-n-butylthiophene, thiophene, 2-butyl, 2-n-butyl thiophene, ccris 6900, 2-butyl-thiophene, thiophene, butyl, acmc-20c3ak
73818
2-butylthiophene
CCCCC1=CC=CS1

2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butylthiophene is an inhibitor of DMH-induced aberrant crypt formation. It is used as a Glutathione S-transferase inducer. It is used as a primary and secondary intermediate.

Solubility
Soluble in acetone and DMSO.

Notes
Storage temperature: 2 - 4°Cand Keep container tightlyclosed in a dryand well-ventilated place. Incompatible to Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.953
Boiling Point 181°C to 182°C
Flash Point 56°C (133°F)
Odor Recognizable
Refractive Index 1.502
Quantity 5 g
UN Number UN1993
Beilstein 107046
Solubility Information Soluble in acetone and DMSO.
Formula Weight 140.25
Percent Purity ≥98%
Chemical Name or Material 2-n-Butylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'