missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(n-Butyl-d9-amino)-4-chloro-6-cyclopropylamino-1,3,5-triazine, TRC

Product Code. 30329107
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30329107 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30329107 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB69034225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(n-Butyl-d9-amino)-4-chloro-6-cyclopropylamino-1,3,5-triaz
CAS 1189997-37-2
Molekylær formel C10 2H9 H7 Cl N5
Mængde 25 mg
InChI formel InChI=1S/C10H16ClN5/c1-2-3-6-12-9-14-8(11)15-10(16-9)13-7-4-5-7/h7H,2-6H2,1H3,(H2,12,13,14,15,16)/i1D3,2D2,3D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC2CC2)n1
IUPAC navn 6-chloro-2-N-cyclopropyl-4-N-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 250.78
Formel vægt 250.1659
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.