Få mere at vide
2-(N-Benzyl-N-methyl)amino-2-phenylbutanol-d5, TRC
Artikelnummer. 30344107
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30344107 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344107 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28566225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(N-Benzyl-N-methyl)amino-2-phenylbutanol-d5
CAS 1330265-76-3
Molekylær formel C18H18D5NO
Mængde 25 mg
Synonym N-Benzyl-N-methyl 1-Phenyl-1-hydroxymethyl-1-propanamine-d5, beta-Ethyl-beta-[methyl(phenylmethyl)amino]benzeneethanol-d5
InChI formel InChI=1S/C18H23NO/c1-3-18(15-20,17-12-8-5-9-13-17)19(2)14-16-10-6-4-7-11-16/h4-13,20H,3,14-15H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(CO)(N(C)Cc1ccccc1)c2ccccc2
IUPAC navn 2-[benzyl(methyl)amino]-3,3,4,4,4-pentadeuterio-2-phenylbutan-1-ol
Molekylvægt (g/mol) 274.41
Formel vægt 274.2093