missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3586675
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1011-41-2
C8H10N2O2S
198.24
MFCD01568825
VDZWHWVAMDQEBT-UHFFFAOYSA-N
2-morpholino-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-thiazole-5-carbaldehyde, 2-morpholinothiazole-5-carbaldehyde, acmc-20anpj, morpholinothiazolecarbaldehyde, 5-formyl-2-morpholin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,2-4-morpholinyl, 2-4-morpholinyl-1,3-thiazole-5-carbaldehyde
1479770
2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde
O=CC1=CN=C(S1)N1CCOCC1
This item is not returnable. View return policy
1011-41-2
C8H10N2O2S
198.24
MFCD01568825
VDZWHWVAMDQEBT-UHFFFAOYSA-N
2-morpholino-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-thiazole-5-carbaldehyde, 2-morpholinothiazole-5-carbaldehyde, acmc-20anpj, morpholinothiazolecarbaldehyde, 5-formyl-2-morpholin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,2-4-morpholinyl, 2-4-morpholinyl-1,3-thiazole-5-carbaldehyde
1479770
2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde
O=CC1=CN=C(S1)N1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 198.25
Percent Purity 97%
Chemical Name or Material 2-Morpholino-1, 3-thiazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'