Få mere at vide
2-Methyltryptamine-d7, TRC
Artikelnummer. 30469783
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30469783 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469783 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33193250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyltryptamine-d7
CAS 1794753-05-1
Molekylær formel C11H7D7N2
Mængde 50 mg
Synonym 2-Methyl-1H-indole-3-ethanamine-d7, 3-(2-Aminoethyl)-2-methyl-indole-d7
InChI formel InChI=1S/C11H14N2/c1-8-9(6-7-12)10-4-2-3-5-11(10)13-8/h2-5,13H,6-7,12H2,1H3/i1D3,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c2c(CCN)c([nH]c2c1[2H])C([2H])([2H])[2H]
IUPAC navn 2-[4,5,6,7-tetradeuterio-2-(trideuteriomethyl)-1H-indol-3-yl]ethanamine
Molekylvægt (g/mol) 181.29
Formel vægt 181.1596