Få mere at vide
2-Methylthiophenothiazine, Mikromol™
Artikelnummer. 30051908
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051908 25 mg
30051315 100 mg
30464061 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051908 Leverandør Mikromol Leverandørnr. MM0116.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methylthiophenothiazine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylthiophenothiazine
CAS 8-5-7643
Analyt- eller komponentnavne 2-Methylthiophenothiazine
Forsendelsestilstand Room Temperature
Molekylær formel C13 H11 N S2
Certificeringer/compliance ISO 17025
Synonym 2-Methylthiophenothiazine
InChI formel InChI=1S/C13H11NS2/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3
SMIL CSc1ccc2Sc3ccccc3Nc2c1
IUPAC navn 2-methylsulfanyl-10H-phenothiazine
Molekylvægt (g/mol) 245.36
Mængde 25 mg
Formel vægt 245.0333 g/mol