missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(Methylthio)benzothiazole, TRC

Artikelnummer. 30480295
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30480295 250 mg -
30480294 500 mg -
30470514 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30480295 Leverandør Toronto Research Chemicals Leverandørnr. TRCM725628250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-22-5
Molekylær formel C8 H7 N S2
Molekylvægt (g/mol) 181.28
Synonym Benzothiazole, 2-(methylthio)- (6CI, 7CI, 8CI, 9CI, ACI), 2-(Methylthio)benzothiazole (ACI), 2-(Methylmercapto)benzothiazole, 2-(Methylthio)benzo[d]thiazole, 2-Methylsulfanyl-1,3-benzothiazole, Methylcaptax, NSC 31259, NSC 41046, NSC 5352
IUPAC navn 2-methylsulfanyl-1,3-benzothiazole
SMIL CSc1nc2ccccc2s1

Specifications

Kemisk navn eller materiale 2-(Methylthio)benzothiazole
Mængde 250 mg
InChI formel InChI=1S/C8H7NS2/c1-10-8-9-6-4-2-3-5-7(6)11-8/h2-5H,1H3
Formel vægt 181.002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.