Få mere at vide
2-Methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-Dioxide, Mikromol™
Artikelnummer. 30052006
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052006 25 mg
30049650 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052006 Leverandør Mikromol Leverandørnr. MM0247.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-Dioxide
CAS 94040-09-2
Analyt- eller komponentnavne 2-Methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C6 H5 N O3 S2
Certificeringer/compliance ISO 17025
Synonym 2-Methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-Dioxide, Tenoxicam Imp. E (EP), Tenoxicam Impurity E
InChI formel InChI=1S/C6H5NO3S2/c1-7-6(8)5-4(2-3-11-5)12(7,9)10/h2-3H,1H3
SMIL CN1C(=O)c2sccc2S1(=O)=O
IUPAC navn 2-methyl-1,1-dioxothieno[2,3-d][1,2]thiazol-3-one
Molekylvægt (g/mol) 203.24
Mængde 25 mg
Formel vægt 202.9711 g/mol