Få mere at vide
2-Methylthiazolidine-4-carboxylic Acid, TRC
Artikelnummer. 30479163
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479163 250 mg
30464838 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479163 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330300250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4165-32-6
Molekylær formel C5H9NO2S
Molekylvægt (g/mol) 147.2
Synonym 2-Methyl-4-thiazolidinecarboxylic Acid
IUPAC navn 2-methyl-1,3-thiazolidine-4-carboxylic acid
SMIL CC1NC(CS1)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Methylthiazolidine-4-carboxylic Acid
Mængde 250 mg
InChI formel InChI=1S/C5H9NO2S/c1-3-6-4(2-9-3)5(7)8/h3-4,6H,2H2,1H3,(H,7,8)
Formel vægt 147.0354