Få mere at vide
2-Methylpyrrole, TRC
Artikelnummer. 30471910
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471910 250 mg
30465609 500 mg
30473619 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471910 Leverandør Toronto Research Chemicals Leverandørnr. TRCM339093250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 636-41-9
Molekylær formel C5H7N
Molekylvægt (g/mol) 81.12
Synonym 2-Methyl-1H-pyrrole, NSC 81346, alpha-Methylpyrrole
IUPAC navn 2-methyl-1H-pyrrole
SMIL Cc1ccc[nH]1
Tekniske data
Kemisk navn eller materiale 2-Methylpyrrole
Mængde 250 mg
InChI formel InChI=1S/C5H7N/c1-5-3-2-4-6-5/h2-4,6H,1H3
Formel vægt 81.0578