Få mere at vide
2-Methylpyridine-d7, CDN
Artikelnummer. 30324196
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324196 1 g -
30322031 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324196 Leverandør CDN Leverandørnr. CDND23201G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Picoline-d7
CAS 93951-93-0
Molekylær formel C6 D7 N
Mængde 1 g
Synonym 2-Methylpyridine-d7, o-Picoline-d7, o-Methylpyridine-d7, ortho-Picoline-d7, ortho-Methylpyridine-d7
InChI formel InChI=1 S/C6H7N/c1-6-4-2-3-5-7-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1nc(c([2 H])c([2 H])c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)pyridine
Molekylvægt (g/mol) 100.17
Formel vægt 100.1018 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity