Få mere at vide
2-Methylpyrazine-d6, TRC
Artikelnummer. 30467487
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30467487 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467487 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3248965MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylpyrazine-d6
CAS 1219804-84-8
Molekylær formel C5 D6 N2
Mængde 5 mg
Synonym 2-Methylpyrazine-d6, 2,3,5-Trideuterio-6-(trideuteriomethyl)pyrazine, 2-(Methyl-d3)pyrazine-3,5,6-d3, 2-(Methyl-d3)-1,4-diazine-3,5,6-d3, 2-Methylpyrazine-D6
InChI formel InChI=1S/C5H6N2/c1-5-4-6-2-3-7-5/h2-4H,1H3/i1D3,2D,3D,4D
SMIL [2H]c1nc([2H])c(nc1[2H])C([2H])([2H])[2H]
IUPAC navn 2,3,5-trideuterio-6-(trideuteriomethyl)pyrazine
Molekylvægt (g/mol) 100.1515
Formel vægt 100.0908