Få mere at vide
2-Methylpyrazine-d6, CDN
Artikelnummer. 30318273
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318273 0.1 g 0.1g
30311725 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318273 Leverandør CDN Leverandørnr. CDND72460.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylpyrazine-d6
CAS 1219804-84-8
Molekylær formel C5 D6 N2
Mængde 0.1 g
Synonym 2-Methylpyrazine-d6, 2,3,5-Trideuterio-6-(trideuteriomethyl)pyrazine, 2-(Methyl-d3)pyrazine-3,5,6-d3, 2-(Methyl-d3)-1,4-diazine-3,5,6-d3, 2-Methylpyrazine-D6
InChI formel InChI=1 S/C5H6N2/c1-5-4-6-2-3-7-5/h2-4 H,1H3/i1D3,2 D,3 D,4 D
SMIL [2 H]c1nc([2 H])c(nc1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,5-trideuterio-6-(trideuteriomethyl)pyrazine
Molekylvægt (g/mol) 100.1515
Formel vægt 100.0908 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity