Få mere at vide
2-Methylpropyl-d9-amine HCl, CDN
Artikelnummer. 30317977
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317977 0.1 g
30310595 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317977 Leverandør CDN Leverandørnr. CDND59630.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isobutylamine-D9 Hydrochloride
CAS 1219799-03-7
Molekylær formel C4 D9 H2 N . Cl H
Mængde 0.1 g
Synonym 1-Propan-1,1,2,3,3,3-d6-amine, 2-(methyl-d3)-, hydrochloride (1:1) (ACI), 2-(Methyl-d3)-1-propan-1,1,2,3,3,3-d6-amine hydrochloride, 2-Methylpropyl-d9-amine hydrochloride, Isobutylamine-d9 hydrochloride, Isobutylamine-D9 Hydrochloride
InChI formel InChI=1 S/C4H11N.ClH/c1-4(2)3-5;/h4H,3,5H2,1-2H3;1 H/i1D3,2D3,3D2,4 D;
SMIL Cl.[2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])N
IUPAC navn 1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 118.6532
Formel vægt 118.1223 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity