Få mere at vide
2-Methylpropyl-d9-amine, CDN
Artikelnummer. 30317001
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317001 0.1 g 0.1g
30322264 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317001 Leverandør CDN Leverandørnr. CDND59380.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylpropyl-d9-amine
CAS 1146967-64-7
Molekylær formel C4 2H9 H2 N
Mængde 0.1 g
InChI formel InChI=1 S/C4H11N/c1-4(2)3-5/h4H,3,5H2,1-2H3/i1D3,2D3,3D2,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])N
IUPAC navn 1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-1-amine
Molekylvægt (g/mol) 82.19
Formel vægt 82.1456 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity