Få mere at vide
2-Methylpropyl Alcohol-OD, CDN
Artikelnummer. 30323226
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30323226 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323226 Leverandør CDN Leverandørnr. CDND72415G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-1-propanol-O-D
CAS 14848-63-6
Molekylær formel C4 D H9 O
Mængde 5 g
Synonym 1-Propanol-d, 2-methyl- (9 CI), Isobutyl alcohol-d (6 CI, 7 CI, 8 CI), 2-Methylpropyl Alcohol-OD, 2-Methylpropan-1-ol-d, Isobutanol-O-d, 2-Methyl-1-propanol-O-D
InChI formel InChI=1 S/C4H10O/c1-4(2)3-5/h4-5 H,3H2,1-2H3/i5D
SMIL [2 H]OCC(C)C
IUPAC navn 1-deuteriooxy-2-methylpropane
Molekylvægt (g/mol) 75.1278
Formel vægt 75.0794 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity