Få mere at vide
(±)-2'-Methylpropranolol Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol Hydrochloride |
| CAS | 15230-34-9 |
| Molekylær formel | C17 H23 N O2 . Cl H |
| Mængde | 1 g |
| Synonym | (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol hydrochloride, Nadolol Imp. F (EP) as Hydrochloride, 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-(1-naphthalenyloxy)-, hydrochloride (1:1), 2-Propanol, 1-(tert-butylamino)-3-(1-naphthyloxy)-, hydrochloride (8CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-(1-naphthalenyloxy)-, hydrochloride (9CI), 1-tert.-Butylamino-3-(1-naphthoxy)-2-propanol hydrochloride, 1-[(1,1-Dimethylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol hydrochloride, (RS)-2'-Methylpropranolol hydrochloride, (±)-2'-Methylpropranolol hydrochloride |
| InChI formel | InChI=1S/C17H23NO2.ClH/c1-17(2,3)18-11-14(19)12-20-16-10-6-8-13-7-4-5-9-15(13)16;/h4-10,14,18-19H,11-12H2,1-3H3;1H |
| SMIL | Cl.CC(C)(C)NCC(O)COc1cccc2ccccc12 |
| IUPAC navn | 1-(tert-butylamino)-3-naphthalen-1-yloxypropan-2-ol;hydrochloride |
| Molekylvægt (g/mol) | 309.83 |
| Formel vægt | 309.1496 |