missing translation for 'onlineSavingsMsg'
Learn More

(2-Methylphenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3175585
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
92614-55-6
C8H9ClO2S
204.668
MFCD02683108
NDMREUGSHNSYBI-UHFFFAOYSA-N
2-methylphenyl methanesulfonyl chloride, o-tolylmethanesulfonyl chloride, o-tolyl-methanesulfonyl chloride, o-tolyl methanesulfonyl chloride, o-tolylmethanesulfonylchloride, 2-methylbenzylsulphonyl chloride, 2-methylphenyl methanesulphonyl chloride, 2-methylphenyl methyl sulfonyl chloride, benzenemethanesulfonyl chloride, 2-methyl, chloro 2-methylphenyl methyl sulfone
2760661
(2-methylphenyl)methanesulfonyl chloride
CC1=CC=CC=C1CS(=O)(=O)Cl
This item is not returnable. View return policy
92614-55-6
C8H9ClO2S
204.668
MFCD02683108
NDMREUGSHNSYBI-UHFFFAOYSA-N
2-methylphenyl methanesulfonyl chloride, o-tolylmethanesulfonyl chloride, o-tolyl-methanesulfonyl chloride, o-tolyl methanesulfonyl chloride, o-tolylmethanesulfonylchloride, 2-methylbenzylsulphonyl chloride, 2-methylphenyl methanesulphonyl chloride, 2-methylphenyl methyl sulfonyl chloride, benzenemethanesulfonyl chloride, 2-methyl, chloro 2-methylphenyl methyl sulfone
2760661
(2-methylphenyl)methanesulfonyl chloride
CC1=CC=CC=C1CS(=O)(=O)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material (2-Methylphenyl)methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'