missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentan-d14, CDN

Product Code. 30318484
Change view
Click to view available options
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30318484 0.5 g
30314220 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318484 Supplier CDN Supplier No. CDND54350.5G
 Begrænset vare
Forventet forsendelse: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Opdag 4000+ højkvalitets deutererede forbindelser, der sendes direkte fra lager for hurtighed. Vi tilbyder mange unikke API'er, herunder aktive farmaceutiske ingredienser, nitrosaminer, aminosyrer, steroider, gasser, pesticider og fedtsyrer.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Methylpentane-D14
CAS 284487-65-6
Molekylær formel C6 D14
Mængde 0.5 g
Synonym Pentane-1,1,1,2,2,3,3,4,5,5,5-d11, 4-(methyl-d3)- (9 CI), 4-(Methyl-d3)pentane-1,1,1,2,2,3,3,4,5,5,5-d11, 2-Methylpentane-d14, 2-Methylpentane-D14
InChI formel InChI=1 S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,3,4,5,5,undecadeuterio-4-(trideuteriomethyl)pentan
Molekylvægt (g/mol) 100.2616
Formel vægt 100,1974 g/mol
Procent renhed 98 atom % D, min 98% kemisk renhed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.