Få mere at vide
2-Methylnaphthalene-D10, TRC
Artikelnummer. 30476633
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30476633 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476633 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32313325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylnaphthalene-d10
CAS 7297-45-2
Molekylær formel C11 2H10
Mængde 25 mg
Synonym Naphthalene-1,2,3,4,5,6,8-d7','7-(methyl-d3)-, 7-(Methyl-d3)naphthalene-1,2,3,4,5,6,8-d7, β-Methylnaphthalene-d10, 2-Methylnaphthalene D10
InChI formel InChI=1S/C11H10/c1-9-6-7-10-4-2-3-5-11(10)8-9/h2-8H,1H3/i1D3,2D,3D,4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c2c([2H])c(c([2H])c([2H])c2c1[2H])C([2H])([2H])[2H]
IUPAC navn 1,2,3,4,5,6,8-heptadeuterio-7-(trideuteriomethyl)naphthalene
Molekylvægt (g/mol) 152.26
Formel vægt 152.141