Få mere at vide
2-Methylnaphthalene-d10, CDN
Artikelnummer. 30322271
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322271 1 g
30316811 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322271 Leverandør CDN Leverandørnr. CDND03541G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylnaphthalene-d10
CAS 7297-45-2
Molekylær formel C11 2H10
Mængde 1 g
Synonym Naphthalene-1,2,3,4,5,6,8-d7, 7-(methyl-d3)-;7-(Methyl-d3)naphthalene-1,2,3,4,5,6,8-d7, β-Methylnaphthalene-d10, 2-Methylnaphthalene D10
InChI formel InChI=1 S/C11H10/c1-9-6-7-10-4-2-3-5-11(10)8-9/h2-8 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c2c([2 H])c(c([2 H])c([2 H])c2c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,3,4,5,6,8-heptadeuterio-7-(trideuteriomethyl)naphthalene
Molekylvægt (g/mol) 152.26
Formel vægt 152.141 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity