Learn More
2-Methylnaphth[1,2-d]oxazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methylnaphth[1,2-d]oxazole |
| CAS | 85-15-4 |
| Molecular Formula | C12 H9 N O |
| Quantity | 1 g |
| Synonym | 2-Methyl-4,5-benzobenzoxazole, 2-Methylnaphtho[1,2-d][1,3]oxazole, 2-Methylnaphtho[1,2-d]oxazole |
| InChI Formula | InChI=1S/C12H9NO/c1-8-13-12-10-5-3-2-4-9(10)6-7-11(12)14-8/h2-7H,1H3 |
| SMILES | Cc1oc2ccc3ccccc3c2n1 |
| IUPAC Name | 2-methylbenzo[e][1,3]benzoxazole |
| Molecular Weight (g/mol) | 183.206 |
| Formula Weight | 183.068 |