Få mere at vide
2-Methylindoline-d3, TRC
Artikelnummer. 30469977
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30469977 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469977 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31349225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylindoline-d3
CAS 1246815-31-5
Molekylær formel C9H8D3N
Mængde 25 mg
Synonym 2,3-Dihydro-2-methyl-1H-indole-d3, NSC 65598-d3, alpha-Methyldihydroindole-d3
InChI formel InChI=1S/C9H11N/c1-7-6-8-4-2-3-5-9(8)10-7/h2-5,7,10H,6H2,1H3/i1D3
SMIL [2H]C([2H])([2H])C1Cc2ccccc2N1
IUPAC navn 2-(trideuteriomethyl)-2,3-dihydro-1H-indole
Molekylvægt (g/mol) 136.21
Formel vægt 136.108