Få mere at vide
2-Methylimidazole-d5 (Major), TRC
Artikelnummer. 30482289
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482289 10 mg
30483032 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482289 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31298710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 697806-98-7
Molekylær formel C4HD5N2
Molekylvægt (g/mol) 87.13
Synonym 1H-2-Methylimidazole-d5, 2-Methyl-1H-imidazole-d5, 2MZ-d5, 2MZ-H-d5, 2MZ-PW-d5, Actiron 2MI-d5, Curezol 2MZ-d5, Curezol 2MZ-P-d5, Denka CN 25-d5, Epicure MI 2-d5, Epikure MI 2-d5, Imicure AMI 2-d5, NSC 21394-d5
SMIL [2H]C1=C(N=C(N1)C([2H])([2H])[2H])[2H]
Tekniske data
Kemisk navn eller materiale 2-Methylimidazole-d5
Mængde 10 mg
InChI formel InChI=1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)/i1D3,2D,3D