Få mere at vide
2-Methylfuran-d6, CDN
Artikelnummer. 30312079
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312079 0.1 g
30324518 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312079 Leverandør CDN Leverandørnr. CDND75120.1G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylfuran-d6
CAS 1398065-93-4
Molekylær formel C5D6O
Mængde 0.1 g
Synonym 5-Methylfuran-d6, NSC 3707-d6, NSC 5211-d6, Silvan-d6, Sylvan-d6, α-Methylfuran-d6;
InChI formel InChI=1 S/C5H6O/c1-5-3-2-4-6-5/h2-4 H,1H3/i1D3,2 D,3 D,4 D
SMIL [2 H]c1oc(c([2 H])c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,4-trideuterio-5-(trideuteriomethyl)furan
Molekylvægt (g/mol) 88.14
Formel vægt 88.0795 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity