Få mere at vide
2-Methyldibenzofuran, TRC
Artikelnummer. 30132974
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132974 250 mg
30010697 500 mg
30012767 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132974 Leverandør Toronto Research Chemicals Leverandørnr. TRCM772240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7320-51-6
Molekylær formel C13 H10 O
Molekylvægt (g/mol) 182.22
Synonym 2-Methyldibenzofuran, 2-Methyldiphenylene oxide
IUPAC navn 2-methyldibenzofuran
SMIL Cc1ccc2oc3ccccc3c2c1
Tekniske data
Kemisk navn eller materiale 2-Methyldibenzofuran
Mængde 250 mg
InChI formel InChI=1S/C13H10O/c1-9-6-7-13-11(8-9)10-4-2-3-5-12(10)14-13/h2-8H,1H3
Formel vægt 182.0732