Få mere at vide
2-Methylbutane-d12, CDN
Artikelnummer. 30317142
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30317142 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317142 Leverandør CDN Leverandørnr. CDND06301G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylbutane-D12
CAS 13351-96-7
Molekylær formel C5 D12
Mængde 1 g
Synonym Butane-1,1,1,2,2,3,4,4,4-d9, 3-(methyl-d3)- (8 CI, 9 CI), Butane-1,1,1,2,3,3,4,4,4-d9, 2-methyl-d3- (7 CI), Isopentane-d12, 3-(Methyl-d3)butane-1,1,1,2,2,3,4,4,4-d9, 2-Methylbutane-d12, 2-Methylbutane-D12
InChI formel InChI=1 S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3/i1D3,2D3,3D3,4D2,5 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,4,4,4-nonadeuterio-3-(trideuteriomethyl)butane
Molekylvægt (g/mol) 84.2227
Formel vægt 84.1692 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity