missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

Catalog Number p-7111781
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
78-78-4
C5H12
72.15
MFCD00009338
QWTDNUCVQCZILF-UHFFFAOYSA-N
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
6556
CHEBI:30362
2-methylbutane
CCC(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
78-78-4
C5H12
72.15
MFCD00009338
QWTDNUCVQCZILF-UHFFFAOYSA-N
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
6556
CHEBI:30362
2-methylbutane
CCC(C)C

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Absorbance max. 0.01 at 400 nm, max. 0.01 at 240 nm, max. 0.07 at 220 nm, max. 0.30 at 210 nm, max. 1.0 at 192 nm
CAS Max % 100.0000%
Density 0.62 g/mL at 25 °C(lit.)
Boiling Point 28°C
Flash Point −51°C
Packaging Glass Bottle
Linear Formula CH3CH2CH(CH3)2
Quantity 1 L
UN Number UN1265
Beilstein 1730723
Formula Weight 72.15
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material 2-Methylbutane, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'