Få mere at vide
2-Methylbutanal 2,4-Dinitrophenylhydrazone, TRC
Artikelnummer. 30472775
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472775 100 mg
30472585 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30472775 Leverandør Toronto Research Chemicals Leverandørnr. TRCM266210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1646-99-7
Molekylær formel C11 H14 N4 O4
Molekylvægt (g/mol) 266.25
Synonym 2-Methylbutanal 2-(2,4-Dinitrophenyl)hydrazone, 2-Methylbutyraldehyde (2,4-Dinitrophenyl)hydrazone, α-Methylbutyraldehyde 2,4-Dinitrophenylhydrazone
IUPAC navn N-(2-methylbutylideneamino)-2,4-dinitro-aniline
SMIL CCC(C)C=NNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Methylbutanal 2,4-Dinitrophenylhydrazone
Mængde 100 mg
InChI formel InChI=1S/C11H14N4O4/c1-3-8(2)7-12-13-10-5-4-9(14(16)17)6-11(10)15(18)19/h4-8,13H,3H2,1-2H3
Formel vægt 266.1015