Få mere at vide
2-Methyl-propanoic-3,3,3-d3 Acid, TRC
Artikelnummer. 30240950
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240950 50 mg
30240866 25 mg
30240937 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240950 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34410050MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 95926-99-1
Molekylær formel C4 D3 H5 O2
Molekylvægt (g/mol) 91.124
Synonym Propanoic-3,3,3-d3 acid, 2-methyl- (9CI), 2-Methylpropanoic-3,3,3-d3 acid, Isobutyric-3,3,3-d6 acid, 2-Methylpropanoic Acid-3,3,3-D3, Isobutyric acid D3 (methyl-D3), Isobutyric acid-d3, 2-Methyl-d3-propionic Acid
IUPAC navn 3,3,3-trideuterio-2-methylpropanoic acid
SMIL [2H]C([2H])([2H])C(C)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Methyl-propanoic-3,3,3-d3 Acid
Mængde 50 mg
InChI formel InChI=1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i1D3
Formel vægt 91.071