Få mere at vide
2-Methyl-N-[3-(trifluoromethyl)phenyl]propanamide, Mikromol™
Artikelnummer. 30049459
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049459 100 mg
30051244 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049459 Leverandør Mikromol Leverandørnr. MM0161.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methyl-N-[3-(trifluoromethyl)phenyl]propanamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-N-[3-(trifluoromethyl)phenyl]propanamide
CAS 1939-27-1
Analyt- eller komponentnavne 2-Methyl-N-[3-(trifluoromethyl)phenyl]propanamide
Forsendelsestilstand Room Temperature
Molekylær formel C11 H12 F3 N O
Certificeringer/compliance ISO 17025
Synonym Flutamide Imp. E (EP), 2-Methyl-N-[3-(trifluoromethyl)phenyl]propanamide, Flutamide Impurity E
InChI formel InChI=1S/C11H12F3NO/c1-7(2)10(16)15-9-5-3-4-8(6-9)11(12,13)14/h3-7H,1-2H3,(H,15,16)
SMIL CC(C)C(=O)Nc1cccc(c1)C(F)(F)F
IUPAC navn 2-methyl-N-[3-(trifluoromethyl)phenyl]propanamide
Molekylvægt (g/mol) 231.21
Mængde 100 mg
Formel vægt 231.0871 g/mol