Få mere at vide
(-)-2-Methyl Isoborneol-d3, TRC
Artikelnummer. 30484092
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30484092 2.5 mg 2.5mg
30470959 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484092 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3140022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 135441-89-3
Molekylær formel C11 2H3 H17 O
Molekylvægt (g/mol) 171.29
Synonym (1R,2R,4R)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol-d3, (1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol-d3, (-)-2-exo-Hydroxy-2-methylbornane-d3
IUPAC navn (1R,3R,4R)-4,7,7-trimethyl-3-(trideuteriomethyl)bicyclo[2.2.1]heptan-3-ol
SMIL [2H]C([2H])([2H])[C@@]1(O)C[C@H]2CC[C@]1(C)C2(C)C
Tekniske data
Kemisk navn eller materiale (-)-2-Methyl Isoborneol-d3
Mængde 2.5 mg
InChI formel InChI=1S/C11H20O/c1-9(2)8-5-6-10(9,3)11(4,12)7-8/h8,12H,5-7H2,1-4H3/t8-,10-,11-/m1/s1/i4D3
Formel vægt 171.1702