Få mere at vide
2-Methyl Hippuric Acid-d7, TRC
Artikelnummer. 30471140
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471140 5 mg
30479622 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471140 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3119835MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216430-90-8
Molekylær formel C10 D7 H4 N O3
Molekylvægt (g/mol) 200.242
Synonym 2-[[2,3,4,5-Tetradeuterio-6-(trideuteriomethyl)benzoyl]amino]acetic acid, N-(2-Methylbenzoyl)glycine-D7 (methylbenzoyl-D7), N-(2-Methylbenzoyl)glycine-d7, 2-Methylhippuric acid-d7, 2-[[2-(Methyl-d3)benzoyl-3,4,5,6-d4]amino]acetic acid, o-Methylhippuric acid-d7, o-Toluric acid-d7, 2-Methyl hippuric acid-d7
IUPAC navn 2-[[2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzoyl]amino]acetic acid
SMIL [2H]c1c([2H])c([2H])c(c(C(=O)NCC(=O)O)c1[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale 2-Methyl Hippuric Acid-d7
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)/i1D3,2D,3D,4D,5D
Formel vægt 200.118
Procent renhed >95%