Få mere at vide
2-Methyl Hippuric Acid-d2, TRC
Artikelnummer. 30482109
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30482109 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482109 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3119821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl Hippuric Acid-d2
CAS 1185100-98-4
Molekylær formel C10 D2 H9 N O3
Mængde 1 mg
Synonym 2,2-Dideuterio-2-[(2-methylbenzoyl)amino]acetic acid, N-(2-Methylbenzoyl)glycine-2,2-D2, N-(2-Methylbenzoyl)glycine-d2, 2-Methylhippuric acid-d2, 2-[(2-Methylbenzoyl)amino]acetic-2,2-d2 acid, o-Methylhippuric acid-d2, o-Toluric acid-d2, 2-Methyl hippuric acid-d2
InChI formel InChI=1S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)/i6D2
SMIL [2H]C([2H])(NC(=O)c1ccccc1C)C(=O)O
IUPAC navn 2,2-dideuterio-2-[(2-methylbenzoyl)amino]acetic acid
Molekylvægt (g/mol) 195.212
Formel vægt 195.086